Skip to Content
Merck
All Photos(1)

Key Documents

G8279

Sigma-Aldrich

Gum guaiac

crude colorimetric peroxidase substrate

Synonym(s):

Guaiac, Guaiacum, Resin guaiac

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201

form

powder

mp

85 °C (dec.) (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Gum guaiac is a tree resin and crude source of 2,5-di-(4-hydroxy-3-methoxyphenyl)-3,4-dimethylfuran (α-guaiaconic acid).[1] The phenolic α-guaiaconic acid acts as a colorimetric peroxidase substrate.[2][3][4] When reacted with an organic or inorganic oxidizing agent, the α-guaiaconic acid is oxidized to a blue-colored quinone. The guaiac reaction has been used to detect trace amounts of heme from hemoglobin in the presence of peroxide.
The Nobles test can be performed using 67% (w/v) gum guaiac diluted in 96% ethanol to analyze the level of extracellular oxidase production [5]

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hermann Brenner et al.
European journal of cancer (Oxford, England : 1990), 49(14), 3049-3054 (2013-05-28)
There is increasing evidence that faecal immunochemical tests (FITs) for haemoglobin offer a number of advantages over traditional guaiac based faecal occult blood tests (gFOBTs). However, evidence on diagnostic performance from direct comparisons with colonoscopy findings in all participants in
Isolation and characterization of α-guaiaconic acid and the nature of guaiacum blue.
Kratochvil, J.F., et al.
Phytochemistry, 10, 2529-2531 (1971)
The Properties and Classification of the Oxidizing Enzymes, and Analogies between Enzymic Activity and the Effects of Immune Bodies and Complements.
E Whitley
The Biochemical journal, 4(3-4), 136-167 (1909-01-01)
C K Palmer et al.
British journal of cancer, 110(7), 1705-1711 (2014-03-13)
Screening for bowel cancer using the guaiac faecal occult blood test offered by the NHS Bowel Cancer Screening Programme (BCSP) is taken up by 54% of the eligible population. Uptake ranges from 35% in the most to 61% in the
S Webendörfer et al.
Deutsche medizinische Wochenschrift (1946), 139(3), 79-83 (2013-10-04)
If the diagnosis is made early the cure rate of bowel cancer is more than 90 %. Occupational preventative medical care required by law and carried out by company physicians can be supplemented by a medical consultation and by simple screenings

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service