Skip to Content
Merck
All Photos(1)

Documents

G7639

Sigma-Aldrich

D-Glucosamine 2,3-disulfate disodium salt

Synonym(s):

2-Deoxy-2-sulfamino-D-glucopyranose 3-sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NO11S2Na2
CAS Number:
Molecular Weight:
383.26
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

[Na].OCC1OC(O)C(NS(O)(=O)=O)C(OS(O)(=O)=O)C1O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service