Skip to Content
Merck
All Photos(1)

Key Documents

G6646

Sigma-Aldrich

Growth Hormone Releasing Factor rat

≥95% (HPLC), powder, synthetic

Synonym(s):

GRF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C225H361N77O66S
CAS Number:
Molecular Weight:
5232.82
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ~70%

UniProt accession no.

storage temp.

−20°C

InChI key

ORNDKVCWLUZAQL-FOWIMTFFSA-N

Gene Information

rat ... Ghrh(29446)

Amino Acid Sequence

His-Ala-Asp-Ala-Ile-Phe-Thr-Ser-Ser-Tyr-Arg-Arg-Ile-Leu-Gly-Gln-Leu-Tyr-Ala-Arg-Lys-Leu-Leu-His-Glu-Ile-Met-Asn-Arg-Gln-Gln-Gly-Glu-Arg-Asn-Gln-Glu-Gln-Arg-Ser-Arg-Phe-Asn

Biochem/physiol Actions

Hypothalamic peptide that stimulates release of somatotropin from anterior pituitary

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Audhya et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(9), 2970-2974 (1985-05-01)
Polyclonal antibodies to synthetic human pancreatic growth hormone-releasing factor [hpGRF(1-44)NH2] and rat hypothalamic growth hormone-releasing factor [rhGRF(1-43)OH] were produced in rabbits by injecting these weak immunogens, coupled to thyroglobulin and emulsified with complete Freund's adjuvant in the presence of activated
N Deslauriers et al.
Regulatory peptides, 20(3), 261-271 (1988-03-01)
We studied the effect of rat growth hormone-releasing factor-(1-43) acid, rGRF(1-43)OH, on the long-term secretion of rat growth hormone (rGH) in dispersed primary cultured cells of rat anterior pituitaries over a period of 7 days or longer. Results of the
J Kraicer et al.
Canadian journal of physiology and pharmacology, 69(2), 181-184 (1991-02-01)
The purpose of this study was to characterize the biological activity of the synthetic rat growth hormone releasing factor analogue rGRF(1-29)NH2 and to compare its action on growth hormone (GH) release to that of authentic rGRF(1-43)OH. We first compared the
L G Frigeri et al.
Endocrinology, 122(6), 2940-2945 (1988-06-01)
Treatment of obese yellow Avy/A mice with the human GH (hGH) peptide hGH-(1-43) enhanced the in vitro sensitivity of their adipose tissue to insulin. Insulin-stimulated glucose oxidation, as determined by measurement of 14CO2 production, was enhanced 106% after administration of
J Spiess et al.
Nature, 303(5917), 532-535 (1983-06-09)
The importance of the hypothalamus in the regulation of growth hormone (GH) secretion from the adenohypophysis has been supported by experimental and clinical evidence. It has been observed that GH secretion is in part controlled by hypothalamic GH release-inhibiting factors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service