Skip to Content
Merck
All Photos(1)

Key Documents

G3157

Sigma-Aldrich

Glucagon

from Porcine pancreas, powder, γ-irradiated, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C153H225N43O49S
CAS Number:
Molecular Weight:
3482.75
MDL number:
UNSPSC Code:
12352207
biological source:
Porcine pancreas
form:
powder
Assay:
≥70% (HPLC)

biological source

Porcine pancreas

Quality Level

sterility

γ-irradiated

Assay

≥70% (HPLC)

form

powder

potency

0.3-10 μg/mL

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl[H].[H]N[C@@H](Cc1cnc[nH]1)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)N[C@@H](C(C)O)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc6c[nH]c7ccccc67)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)O)C(O)=O

InChI

1S/C153H225N43O49S.ClH/c1-72(2)52-97(133(226)176-96(47-51-246-11)132(225)184-104(60-115(159)209)143(236)196-123(78(10)203)151(244)245)179-137(230)103(58-83-64-167-89-29-19-18-28-87(83)89)183-131(224)95(43-46-114(158)208)177-148(241)120(74(5)6)194-141(234)101(54-79-24-14-12-15-25-79)182-138(231)105(61-117(211)212)185-130(223)94(42-45-113(157)207)171-124(217)75(7)170-127(220)91(31-22-49-165-152(160)161)172-128(221)92(32-23-50-166-153(162)163)174-146(239)110(69-199)191-140(233)107(63-119(215)216)186-134(227)98(53-73(3)4)178-135(228)99(56-81-33-37-85(204)38-34-81)180-129(222)90(30-20-21-48-154)173-145(238)109(68-198)190-136(229)100(57-82-35-39-86(205)40-36-82)181-139(232)106(62-118(213)214)187-147(240)111(70-200)192-150(243)122(77(9)202)195-142(235)102(55-80-26-16-13-17-27-80)188-149(242)121(76(8)201)193-116(210)66-168-126(219)93(41-44-112(156)206)175-144(237)108(67-197)189-125(218)88(155)59-84-65-164-71-169-84;/h12-19,24-29,33-40,64-65,71-78,88,90-111,120-123,167,197-205H,20-23,30-32,41-63,66-70,154-155H2,1-11H3,(H2,156,206)(H2,157,207)(H2,158,208)(H2,159,209)(H,164,169)(H,168,219)(H,170,220)(H,171,217)(H,172,221)(H,173,238)(H,174,239)(H,175,237)(H,176,226)(H,177,241)(H,178,228)(H,179,230)(H,180,222)(H,181,232)(H,182,231)(H,183,224)(H,184,225)(H,185,223)(H,186,227)(H,187,240)(H,188,242)(H,189,218)(H,190,229)(H,191,233)(H,192,243)(H,193,210)(H,194,234)(H,195,235)(H,196,236)(H,211,212)(H,213,214)(H,215,216)(H,244,245)(H4,160,161,165)(H4,162,163,166);1H/t75-,76+,77+,78+,88-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,120-,121-,122-,123-;/m0./s1

InChI key

RKGLLHCSSVJTAN-YYICOITRSA-N

Gene Information

pig ... GCG(397595)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr

Biochem/physiol Actions

Glucagon is a 29-amino acid pancreas secreted peptide hormone that raises blood glucose and insulin levels. Glucagon acts via a G protein-coupled glucagon receptor. The glucagon GPCR induces an adenylate cyclase:protein kinase A:phosphorylase kinase:glycogen phosphorylase-A signaling pathway that results in the release of glucose-1-phosphate from glycogen.

Components

H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH.xHCl

Physical form

powder-0 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirstan A Vessey et al.
Investigative ophthalmology & visual science, 46(11), 3922-3931 (2005-10-27)
In chicks, plus defocus retards eye growth, thickens the choroid, and activates glucagonergic amacrine cells, probably releasing glucagon. Glucagon receptor antagonists (expected to inhibit compensation to plus defocus) and agonists (expected to block myopia induction by form deprivation) were administered
Shiva Reddy et al.
Diabetologia, 58(5), 1024-1035 (2015-02-18)
The role of peri-islet CD45-positive leucocytes, as one component of insulitis, in beta cell death during human type 1 diabetes remains unclear. We undertook a case study, comparing and quantifying leucocytes in the peri- and intra-islet areas in insulin-positive and
Nicolai J Wewer Albrechtsen et al.
Diabetologia, 57(9), 1919-1926 (2014-06-04)
Hyperglucagonaemia is a characteristic of several clinical conditions (e.g. end-stage renal disease (ESRD), type 2 diabetes, obesity before and after Roux-en-Y gastric bypass (RYGB) and vagotomy with pyloroplasty), but the molecular nature of 'immunoreactive' glucagon is poorly characterised. The specific
Liehua Liu et al.
Biochemical and biophysical research communications, 453(3), 398-404 (2014-10-01)
Dipeptidyl peptidase 4 (DPP-4) was recently found to be expressed in human and mouse islets with different expression patterns. However, whether species-dependent expression pattern is a generalized phenomenon and whether islet DPP-4 activity is regulated are not known. This study
Bilal A Omar et al.
Diabetologia, 57(9), 1876-1883 (2014-06-19)
Inhibition of the enzyme dipeptidyl peptidase 4 (DPP-4), which cleaves and inactivates glucagon-like peptide 1 (GLP-1), is a glucose-lowering strategy in type 2 diabetes. Since DPP-4 is a ubiquitously distributed enzyme, we examined whether it is expressed in islets and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service