Skip to Content
Merck
All Photos(1)

Key Documents

G1920

Sigma-Aldrich

Anti-Glutamate Receptor 7, Metabotropic antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-mGluR7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using human brain, neurons

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRM7(2917)
mouse ... Grm7(108073)
rat ... Grm7(81672)

General description

The metabotropic glutamate receptors consist of eight subtypes (mGluR1-8) divided into three groups (I-III). Glutamate is the main excitatory neurotransmitter in the brain. It acts on ligand-gated receptor channels, termed NMDA, AMPA and kainate receptors, involved in the fast excitatory synaptic transmission. Glutamate also regulates ion channels and enzymes producing second messengers via specific receptors coupled to G-proteins, called metabotropic glutamate receptors.

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human metabotropic glutamate receptor 7, conjugate to KLH. The immunizing peptide has 100% homology with the rat gene and 95% homology with the mouse gene.

Application

Anti-Glutamate Receptor 7, Metabotropic antibody produced in rabbit is suitable for immunohistochemistry using formalin-fixed, paraffin-embedded sections of human brain, neurons.

Biochem/physiol Actions

Glutamate Receptor 7 belongs to group III and inhibits adenyl cyclase. The role of the metabotropic receptors in pain and pathology is being studied for therapeutic potential. Activation of Group III receptors has been shown to be neuroprotective. Glutamate receptors are important mediators of excitatory amino acid neurotransmission.

Physical form

Solution in phosphate buffered saline, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Faden et al.
Journal of neurotrauma, 14(12), 885-895 (1998-02-25)
We have used an in vitro trauma model to examine the effects of modulation of group III metabotropic glutamate receptors (mGluR) on post-traumatic neuronal cell death. Rat cortical neuronal/glial cultures were subjected to standardized mechanical injury using a punch that
V Bruno et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 20(17), 6413-6420 (2000-08-31)
Activation of group III metabotropic glutamate receptors (mGluR4, mGluR6, mGluR7, and mGluR8) has been established to be neuroprotective in vitro and in vivo. To disclose the identity of the receptor subtype(s) that exert(s) the protective effect, we have used group
W Spooren et al.
Behavioural pharmacology, 14(4), 257-277 (2003-07-03)
Following the molecular cloning in the early 1990s of the metabotropic glutamate receptors (mGlu1-8), research that focused on the physiology, pharmacology and function of these receptors revealed their potential role in CNS disorders. Numerous psychiatric and neurological dis-orders are indeed
J P Pin et al.
Neuropharmacology, 34(1), 1-26 (1995-01-01)
Glutamate is the main excitatory neurotransmitter in the brain. For many years it has been considered to act only on ligand-gated receptor channels--termed NMDA, AMPA and kainate receptors--involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to
Domenico E Pellegrini-Giampietro
Trends in pharmacological sciences, 24(9), 461-470 (2003-09-12)
Metabotropic glutamate receptors of the mGlu(1) and mGlu(5) subtypes exhibit a high degree of sequence homology and are both coupled to phospholipase C and intracellular Ca(2+) mobilization. However, functional differences have been detected for these receptor subtypes when they are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service