Skip to Content
Merck
All Photos(1)

Key Documents

G1794

Sigma-Aldrich

GAP-27

≥97% (HPLC)

Synonym(s):

Ser-Arg-Pro-Thr-Glu-Lys-Thr-Ile-Phe-Ile-Ile

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€559.00

€559.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 MG
€559.00

About This Item

Empirical Formula (Hill Notation):
C60H102N15O17
CAS Number:
Molecular Weight:
1305.54
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€559.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

Assay

≥97% (HPLC)

form

powder

packaging

pkg of 2 mg

technique(s)

cell culture | mammalian: suitable

solubility

water: soluble 1.00-1.02 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

InChI

1S/C60H101N15O17/c1-9-31(4)44(54(86)69-41(29-36-19-13-12-14-20-36)52(84)70-45(32(5)10-2)55(87)72-46(59(91)92)33(6)11-3)71-57(89)48(35(8)78)73-51(83)38(21-15-16-26-61)66-50(82)39(24-25-43(79)80)67-56(88)47(34(7)77)74-53(85)42-23-18-28-75(42)58(90)40(22-17-27-65-60(63)64)68-49(81)37(62)30-76/h12-14,19-20,31-35,37-42,44-48,76-78H,9-11,15-18,21-30,61-62H2,1-8H3,(H,66,82)(H,67,88)(H,68,81)(H,69,86)(H,70,84)(H,71,89)(H,72,87)(H,73,83)(H,74,85)(H,79,80)(H,91,92)(H4,63,64,65)/t31-,32-,33-,34+,35+,37-,38-,39-,40-,41-,42-,44-,45-,46-,47-,48-/m0/s1

InChI key

SXRAPDIXXYFGJG-MDAHIHQXSA-N

Gene Information

human ... GJA4(2701)

Amino Acid Sequence

Ser-Arg-Pro-Thr-Glu-Lys-Thr-Ile-Phe-Ile-Ile

Application

GAP-27 has been used as a blocking peptide for Cx43 (connexin 43) gap junctions in FDC (follicular dendritic cells)-B cell cultures.[1]

Biochem/physiol Actions

GAP-27 shares homology with a region of extracellular loops of connexins, and hence, is thought to inhibit junctions formed by the connexins Cx32, Cx40 and Cx43. It highly hinders co-operative cell-cell interactions including the synergized beating of embryonic cardiomyocytes. This peptide reduces osteoclastic activity and survival rates.[2] This peptide significantly reduces acetylcholine-mediated hyperpolarization of smooth muscle cells.[3]
Inhibitor of gap junction signaling in endothelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaun L Sandow et al.
Circulation research, 90(10), 1108-1113 (2002-06-01)
The nature of the vasodilator endothelium-derived hyperpolarizing factor (EDHF) is controversial, putatively involving diffusible factors and/or electrotonic spread of hyperpolarization generated in the endothelium via myoendothelial gap junctions (MEGJs). In this study, we investigated the relationship between the existence of
Hajnalka Rajnai et al.
Journal of immunology research, 2015, 528098-528098 (2015-03-31)
Follicular dendritic cells (FDC) show homo- and heterocellular metabolic coupling through connexin 43 (Cx43) gap junctions and support B cell selection and maturation in germinal centers. In follicular lymphomas B cells escape apoptosis while FDC develop abnormally. Here we tested
J Ilvesaro et al.
BMC musculoskeletal disorders, 2, 10-10 (2001-12-19)
Bone remodelling is dependent on the balance between bone resorbing osteoclasts and bone forming osteoblasts. We have shown previously that osteoclasts contain gap-junctional protein connexin-43 and that a commonly used gap-junctional inhibitor, heptanol, can inhibit osteoclastic bone resorption. Since heptanol
Wenru Su et al.
Journal of immunology (Baltimore, Md. : 1950), 203(6), 1436-1446 (2019-08-20)
Therapeutic manipulation of regulatory T cells (Tregs) has been regarded as a promising approach for the treatment of immune disorders. However, a better understanding of the immunomodulatory mechanisms of Tregs and new safe and effective methods to improve the therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service