Skip to Content
Merck
All Photos(1)

Key Documents

G11100P

Sigma-Aldrich

DMT-dG(ib) Pharmadite®

Synonym(s):

DMT-dG(ib) Phosphoramidite, N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyguanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 5 G
€824.00

€824.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
6 X 5 G
€824.00

About This Item

Empirical Formula (Hill Notation):
C44H54N7O8P
CAS Number:
Molecular Weight:
839.92
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.51

€824.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99.5% (reversed phase HPLC)
>99.5% (31P-NMR)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

>= 170ppm, none detected (S/N <2.5)
≤0.1% P(III) 100-169ppm (31P-NMR)
≤0.1% single unspecified Imprity (reversed phase HPLC)
≤0.3% dG2 (reversed phase HPLC, DMT-dG(ib) cyanoethyl-H-posphonate)
≤0.3% dG3 (reversed phase HPLC, DMT-dG(ib) phosphoramidate)
≤0.3% dG4 (reversed phase HPLC, DMT-dG(ib)-diisopropylamino-H-phosphonate)
≤0.30 wt. % water content (Karl Fischer)
≤0.5% P(V) -25-99ppm (31P-NMR)
≤3.0 wt. % residual solvent content

color

white to off-white

solubility

soluble, clear

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: deoxyguanosine
base protecting group: isobutyryl
2' protecting group: none
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C44H54N7O8P/c1-28(2)41(52)48-43-47-40-39(42(53)49-43)46-27-50(40)38-25-36(59-60(57-24-12-23-45)51(29(3)4)30(5)6)37(58-38)26-56-44(31-13-10-9-11-14-31,32-15-19-34(54-7)20-16-32)33-17-21-35(55-8)22-18-33/h9-11,13-22,27-30,36-38H,12,24-26H2,1-8H3,(H2,47,48,49,52,53)/t36-,37+,38+,60?/m0/s1

InChI key

FDRMKYVTIFSDPR-MMROLVBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dG(ib) Pharmadite® is a Pharmadite® product line that represents a new class of standard protected DNA phosphoramidites designed with highly controlled impurity profiles and exceptional overall purity. Pharmadite phosphoramidites are characterized by their exceptional purity and well-defined impurity profile. In particular, they are essentially free from contaminants that interfere in coupling reactions, such as other nucleosidic or non-nucleosidic phosphoramidites (P(III)-contaminants). All remaining impurities in Pharmadite phosphopramidites, if present, are identified, characterized, and quantified. The impact of any such defined impurities on the synthesis of oligonucleotides is well understood and has been shown to be insignificant.

Legal Information

Pharmadite is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service