Skip to Content
Merck
All Photos(1)

Key Documents

G0754

Sigma-Aldrich

D-Glucoheptono-1,4-lactone

≥99%

Synonym(s):

α-D-Glucoheptonic γ-lactone, D-Glycero-D-guloheptono-γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O7
CAS Number:
Molecular Weight:
208.17
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99%

mp

152-155 °C (lit.)

SMILES string

[H][C@]1(OC(=O)[C@H](O)[C@@H]1O)[C@H](O)[C@H](O)CO

InChI

1S/C7H12O7/c8-1-2(9)3(10)6-4(11)5(12)7(13)14-6/h2-6,8-12H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

VIVCRCODGMFTFY-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Glucoheptono-1,4-lactone may be used for the organic synthesis of N-alkyl-N-(2-hydroxyethyl)aldonamides and N-cycloalkylaldonamides.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorota Piłakowska-Pietras et al.
Journal of colloid and interface science, 294(2), 423-428 (2005-09-13)
The foamability of the aqueous solutions of functionalized, surface-chemically pure surfactants of the nonionic saccharide-type: N,N-di-n-alkylaldonamides, N-alkyl-N-(2-hydroxyethyl)aldonamides, and N-cycloalkylaldonamides, derivatives of D-glucono-1,5-lactone and/or D-glucoheptono-1,4-lactone, were investigated. The approach of Lunkenheimer and Małysa for the characterization of the foamability and foam
Dorota Piłakowska-Pietras et al.
Langmuir : the ACS journal of surfaces and colloids, 20(5), 1572-1578 (2005-04-02)
N-alkyl-N-(2-hydroxyethyl)aldonamides (alkyl: n-C6H13, n-C8H17, n-C10H21, n-C12H25, and n-C14H29) were obtained in the reaction of long-chain N-alkyl-N-(2-hydroxyethyl)amines with D-glucono-1,5-lactone and D-glucoheptono-1,4-lactone. The adsorption isotherms were obtained from surface tension measurements of aqueous solutions of surface-chemically pure surfactants. The experimental equilibrium surface

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service