Skip to Content
Merck
All Photos(1)

Key Documents

G0545

Sigma-Aldrich

Anti-GABAA Receptor (γ2 subunit) (extracellular) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-γ-Aminobutyric Acid Type A Receptor (γ2 subunit)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€272.00
200 μL
€734.00

€272.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μL
€272.00
200 μL
€734.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€272.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

bovine, mouse, rat, human

technique(s)

western blot: 1:200 using rat brain membrane

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GABRG2(2566)
mouse ... Gabrg2(14406)
rat ... Gabrg2(29709)

Immunogen

synthetic peptide corresponding to amino acids 39-53 of rat GABAA receptor (γ2 subunit). The sequence is identical in human, mouse, and bovine. In chick, it is 13/15 residues identical.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 1% BSA, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina A Oliva et al.
International journal of molecular sciences, 24(2) (2023-01-22)
In Octodon degus, the aging process is not equivalent between sexes and worsens for females. To determine the beginning of detrimental features in females and the ways in which to improve them, we compared adult females (36 months old) and
Ann J Johnston et al.
Neurobiology of disease, 64, 131-141 (2014-01-11)
Genetic mutations in voltage-gated and ligand-gated ion channel genes have been identified in a small number of Mendelian families with genetic generalised epilepsies (GGEs). They are commonly associated with febrile seizures (FS), childhood absence epilepsy (CAE) and particularly with generalised

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service