Skip to Content
Merck
All Photos(3)

Key Documents

G0174

Sigma-Aldrich

Picroside II

Synonym(s):

(1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-[(4-hydroxy-3-methoxybenzoyl)oxy]-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl-β-D-glucopyranoside, 6-Vanillylcatalpol, Amphicoside II, Vanilloyl catalpol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28O13
CAS Number:
Molecular Weight:
512.46
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.25

Assay

≥98% (HPLC)

solubility

ethanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC1[C@@H](O)[C@H](O)C(CO)O[C@H]1O[C@@H]2OC=C[C@]3([H])[C@@]2([H])[C@](O4)(CO)[C@@H]4[C@H]3OC(C5=CC=C(O)C(OC)=C5)=O

InChI

1S/C23H28O13/c1-31-12-6-9(2-3-11(12)26)20(30)34-18-10-4-5-32-21(14(10)23(8-25)19(18)36-23)35-22-17(29)16(28)15(27)13(7-24)33-22/h2-6,10,13-19,21-22,24-29H,7-8H2,1H3

InChI key

AKNILCMFRRDTEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PicrosideII (PII) is a monoterpene, iridoid glycoside, and one of the main bioactivecompounds sourced from Picrorhiza kurroa.

Application

PicrosideII has been used as a reference standard to analyze and validate picroside-II from Picrorhiza kurroa using high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

Phytochemical found in Traditional Chinese Medicine herbal preparations. Picroside II, a glucoside, is reported to have hepatoprotective, cardioprotective, and neuroprotective properties.
Picroside II displaysmany anti-cancer properties such as anti-metastatic, anti-angiogenic, and anti-apoptotic activities in both in vitro and in vivo studies.It also exerts antioxidant, anti-inflammatory, and antidiarrheal effectsagainst acute oxidative injuries such as severe acute pancreatitis-inducedintestinal barrier injury. PII has protective effects onischemia/reperfusion (I/R)-induced injury by interacting with differentmechanisms in various in vitro and in vivo models.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenghua Lou et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 120, 109494-109494 (2019-10-14)
Cancer is one of the leading causes of death worldwide. The development of novel anti-cancer agents from natural products is a promising approach to reduce cancer mortality. In this study, we investigated the anti-metastatic and anti-angiogenic activities of picroside II
Development and Validation of HPLC Method for Analysis of Picroside-I and Picroside-II in Picrorhiza kurroa
Sharma S, et al.,
International Research Journal of Pure and Applied Chemistry, 17, 1-10 (2019)
Shangying Ma et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 130, 110421-110421 (2020-07-17)
Picrorhiza kurroa, the dried rhizome of Picrorhiza kurroa Royle ex Benth, is a famous Chinese herb that has been traditionally used in China. Picroside II (PII), a glycoside derivative, is the main bioactive constituent of Picrorhiza kurroa. In the past
Xuehua Piao et al.
Oxidative medicine and cellular longevity, 2020, 3589497-3589497 (2020-05-01)
Picroside II exerts anti-inflammatory and antidiarrheal effects for treating the diseases associated with oxidative injury. However, its function on pancreatitis-induced intestinal barrier injury remains unclear. Hypothesis/Purpose. We hypothesized that picroside II will have protective effects against pancreatitis-induced intestinal barrier injury

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service