Skip to Content
Merck
All Photos(1)

Key Documents

FFW

Sigma-Aldrich

Fireflies, whole, desiccated

for bioluminescence

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352202

technique(s)

toxicology assay: suitable

storage temp.

−20°C

Application

Fireflies have been used in several studies of bioluminescence. In particular, they may be used in assays to determine toxicity pathways because bioluminescence can quantitatively measure a wide range of biological activities.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara N Kirillova et al.
Analytical and bioanalytical chemistry, 400(2), 343-351 (2011-02-22)
The paper investigates an application of luminescent bioassays to monitor the toxicity of organic halides. Effects of xanthene dyes (fluorescein, eosin Y, and erythrosin B), used as model compounds, on bioluminescent reactions of firefly Luciola mingrelica, marine bacteria Photobacterium leiognathi
Tamara N Kirillova et al.
Analytical and bioanalytical chemistry, 387(6), 2009-2016 (2007-01-24)
Bioluminescent reactions of luminous organisms are excellent models for studying the effects of heavy atoms on enzymatic processes. The effects of potassium halides with halide anions of different atomic weight were compared in bioluminescent reactions of the firefly (Luciola mingrelica)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service