Skip to Content
Merck
All Photos(1)

Key Documents

F8799

Sigma-Aldrich

Fas/Fc Chimera from mouse

>97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

Apo-1, CD95

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€515.00

€515.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€515.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

€515.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

0.03-0.1 μg per mL ED50 (in the presence of 20 ng/mL recombinant human Fas ligand for cytotoxicty assay of Jurkat cells)

mol wt

55 kDa (glycosylated form)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Fas(14102)

General description

Fas is a key component of apoptosis, and is a cell surface receptor. This gene is localized to human chromosome 10q24.1, and is composed of nine exons and eight introns.[1]

Application

Fas/Fc Chimera from mouse has been used as a blocking agent during FACS (flow cytometry).[2]

Biochem/physiol Actions

Fas and its ligand play an essential role in apoptosis, by activating the cell death signaling pathway. Mutations in this gene are associated with evasion of apoptosis by cells, leading to tumorigenesis, and prevention of clearance of these cells by the immune system.[1] A polymorphism in this gene results in the production of soluble Fas (sFas), which is implicated in the chronic autoimmune disorder called primary Sjögren′s syndrome (pSS).[3]

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

Biological activity was measured by its ability to inhibit Fas-ligand induced apoptosis of Jurkat cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davidson D Ateh et al.
Biomaterials, 32(33), 8538-8547 (2011-08-10)
The CD95/CD95L receptor-ligand system is mainly recognised in the induction of apoptosis. However, it has also been shown that CD95L is over-expressed in many cancer types where it modulates immune-evasion and together with its receptor CD95 promotes tumour growth. Here
J Cheng et al.
Science (New York, N.Y.), 263(5154), 1759-1762 (1994-03-25)
Fas is an apoptosis-signaling receptor molecule on the surface of a number of cell types. Molecular cloning and nucleotide sequence analysis revealed a human Fas messenger RNA variant capable of encoding a soluble Fas molecule lacking the transmembrane domain because
Cheng Jin et al.
Medical science monitor : international medical journal of experimental and clinical research, 21, 630-637 (2015-02-28)
The CD95 gene plays a key role in regulating cell growth and tumor genesis. To date, several publications have focused on the CD95 rs1800682A/G site polymorphism and various types of tumors in Asians; however, this association is still controversial and
A Oehm et al.
The Journal of biological chemistry, 267(15), 10709-10715 (1992-05-25)
The APO-1 antigen as defined by the mouse monoclonal antibody anti-APO-1 was previously found to be expressed on the cell surface of activated human T and B lymphocytes and a variety of malignant human lymphoid cell lines. Cross-linking of the
B A Treviño-Talavera et al.
Genetics and molecular research : GMR, 13(3), 4831-4838 (2014-07-26)
Primary Sjögren's syndrome (pSS) is a chronic systemic autoimmune disease characterized by lymphocytic infiltration of exocrine glands. Soluble Fas receptor (sFas) has been suggested as a Fas-mediated apoptosis blocker that could impair clonal deletion in infiltrated autoreactive cells. The FAS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service