Skip to Content
Merck
All Photos(1)

Key Documents

F8381

Sigma-Aldrich

Fructose-6-phosphate Kinase, Pyrophosphate-dependent from Propionibacterium freudenreichii (shermanii)

lyophilized powder, ≥4.0 units/mg protein

Synonym(s):

6-Phosphofructokinase(pyrophosphate), Pyrophosphate: D-fructose-6-phosphate 1-phosphotransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥4.0 units/mg protein

composition

Protein, 5.0-25.0% biuret

storage temp.

−20°C

Application

FBP was used to study the kinetic mechanism of pyrophosphate-dependent phosphofructokinase from Propionibacterium freudenreichii.

Biochem/physiol Actions

Fructose-1,6-bisphosphatase (FBP) is an important enzyme in glucose metabolism. It catalyzes the hydrolysis of fructose-1,6-bisphosphate to fructose-6-phosphate and inorganic phosphate. Fructose-6-phosphate kinase converts fructose-6-phosphate into fructose 1,6-bisphophate in the rate limiting step of the glycolysis cycle.

Other Notes

These two pyrophosphate-dependent fructose-6-phosphate kinases differ in their response to the physiological activator fructose 2,6-diphosphate [Fru (2,6)-P2]. The enzyme from Propionibacterium freudenreichii (F8381) reportedly is not affected by Fru (2,6)-P2. The enzyme from potato tubers (F2258) is stimulated by Fru (2,6)-P2 (half-maximal activation at 5.5 nM Fru (2,6)-P2 for potato tuber enzyme).

Unit Definition

One unit will convert 1.0 μmole of pyrophosphate and fructose 6-phosphate to fructose 1,6-diphosphate and inorganic phosphate per min at pH 7.4 at 30 °C.

Physical form

Contains imidazole salts and stabilizer

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corrigendum: Online Pyrophosphate Assay for Analyzing Adenylation Domains of Nonribosomal Peptide Synthetases.
Tiia Kittilä et al.
Chembiochem : a European journal of chemical biology, 23(18), e202200381-e202200381 (2022-07-28)
Daniel L Machell et al.
Chembiochem : a European journal of chemical biology, 23(18), e202200325-e202200325 (2022-07-26)
Detection of pyrophosphate is important in quantifying enzyme activity, particularly adenylation domain activity during non-ribosomal peptide synthesis. The previous development of an enzyme coupled PPi /NADH assay allowed the measurement of such activity in an online fashion using commercially available
B L Bertagnolli et al.
Biochemistry, 23(18), 4101-4108 (1984-08-28)
Inorganic pyrophosphate dependent D-fructose-6-phosphate 1-phosphotransferase from Propionibacterium freudenreichii was purified to apparent homogeneity by the criterion of silver staining on sodium dodecyl sulfate (SDS) gels. In the direction of phosphorylation of fructose 6-phosphate (F6P), an intersecting initial velocity pattern is
M Müller et al.
Journal of bacteriology, 183(22), 6714-6716 (2001-10-24)
Inorganic pyrophosphate-dependent phosphofructokinase (PP(i)-PFK) of the amitochondriate eukaryote Mastigamoeba balamuthi was sequenced and showed about 60% identity to PP(i)-PFKs from two eubacteria, Propionibacterium freudenreichii and Sinorhizobium meliloti. These gene products represent a newly recognized lineage of PFKs. All four lineages
Florencio E Podestá et al.
Archives of biochemistry and biophysics, 414(1), 101-107 (2003-05-15)
The intrinsic fluorescence of potato tuber pyrophosphate:fructose-6-phosphate 1-phosphotransferase (PFP) was used as an indicator of conformational changes due to ligand binding. Binding of the substrates and the allosteric activator fructose-2,6-bisphosphate was quantitatively compared to their respective kinetic effects on enzymatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service