Skip to Content
Merck
All Photos(1)

Key Documents

F6000

Sigma-Aldrich

[2,2′]-Furildioxime mixed isomers hydrate

Synonym(s):

[2,2’]-Furildioxime monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O4
CAS Number:
Molecular Weight:
220.18
Beilstein:
85894
EC Number:
MDL number:
UNSPSC Code:
12352103

storage temp.

−20°C

SMILES string

O\N=C(c1ccco1)/C(=N\O)c2ccco2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service