Skip to Content
Merck
All Photos(1)

Key Documents

F5804

Sigma-Aldrich

Anti-FBG3 (FBXO44) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-F Box 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa by calculation

species reactivity

human

technique(s)

western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Immunogen

peptide corresponding to the human FBG3 protein (amino acids 103-119). This sequence has 94% homology with mouse and rat.

Application

Anti-FBG3 (FBXO44) antibody is suitable for western blot: 1:500-1:1,000.

Biochem/physiol Actions

F-box protein, is the critical component of conserved ubiquitin–protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box) and acts as a substrate recognition component of the ubiquitin ligase complex SCF (Skp-Cullin-F-box). It is expressed in human tissues including kidney, liver, brain and muscle tissues. It is associated with various critical biological processes such as in regulating the cell cycle, immune response, signaling cascades and developmental programs. The proteins have a bipartite structure and the core complex, whereas divergent protein-protein interaction motifs selectively bind their cognate substrates.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gennady P Ilyin et al.
Gene, 296(1-2), 11-20 (2002-10-18)
F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Here, we report the identification of full-length cDNAs encoding three novel human F-box
K L Craig et al.
Progress in biophysics and molecular biology, 72(3), 299-328 (1999-12-03)
The ubiquitin system of intracellular protein degradation controls the abundance of many critical regulatory proteins. Specificity in the ubiquitin system is determined largely at the level of substrate recognition, a step that is mediated by E3 ubiquitin ligases. Analysis of
Laura Pontano Vaites et al.
Molecular and cellular biology, 31(22), 4513-4523 (2011-09-14)
Skp1-Cul1-F-box (SCF) E3 ubiquitin ligase complexes modulate the accumulation of key cell cycle regulatory proteins. Following the G(1)/S transition, SCF(Fbx4) targets cyclin D1 for proteasomal degradation, a critical event necessary for DNA replication fidelity. Deregulated cyclin D1 drives tumorigenesis, and
Margaret S Ho et al.
Journal of biomedical science, 13(2), 181-191 (2006-02-08)
The eukaryotic protein degradation pathway involves the ubiquitin (Ub) modification of substrates targeted for degradation by the 26S proteasome. The addition of Ub, a process called ubiquitination, is mediated by enzymes including the E3 Ub ligases which transfer the Ub

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service