Skip to Content
Merck
All Photos(1)

Key Documents

F3893

Sigma-Aldrich

Anti-Monkey IgG (whole molecule)−FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Monkey IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€184.00

€184.00


Please contact Customer Service for Availability


Select a Size

Change View
1 ML
€184.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

€184.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

dot immunobinding: 1:64

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.

Specificity

Anti-Monkey IgG (whole molecule) antibody recognizes all monkey IgG subclasses. Rabbit anti-Monkey IgG is conjugated to Fluorescein Isothiocyanate (FITC).The identity and purity of affinity isolated antibody is established by immunoelectrophoresis.

Immunogen

Purified monkey IgG (rhesus)

Application

Anti-Monkey IgG (whole molecule)-FITC antibody produced in rabbit has been used in western blotting and indirect immunofluorescence.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SIVsm Tat, Rev, and Nef1: functional characteristics of r-GV internalization on isotypes, cytokines, and intracellular degradation
Sremac M and Stuart ES
BMC biotechnology, 10(1), 54-54 (2010)
Widespread occurrence of antibodies against circumsporozoite protein and against blood forms of Plasmodium vivax, P. falciparum and P. malariae in Brazilian wild monkeys
Duarte AMRC, et al.
Journal of Medical Primatology, 35(2), 87-96 (2006)
Béatrice Volney et al.
Acta tropica, 82(1), 11-23 (2002-03-21)
This paper describes a sero-epidemiological study of malaria prevalence in French Guiana. An immunofluorescence assay and an enzyme-linked immunosorbent assay were used to detect antibodies against blood-stage antigens and synthetic peptides mimicking the repetitive epitope of the sporozoites of Plasmodium
S Hashira et al.
Pediatrics international : official journal of the Japan Pediatric Society, 42(4), 337-342 (2000-09-15)
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service