F1884
(±)-Fenfluramine hydrochloride solution
drug standard, 1.0 mg/mL in methanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
drug standard
Assay
≥98%
concentration
1.0 mg/mL in methanol
storage temp.
2-8°C
SMILES string
Cl.CCNC(C)Cc1cccc(c1)C(F)(F)F
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service