F1509
Anti-Human Fibronectin antibody produced in goat
fractionated antiserum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
biological source
goat
conjugate
unconjugated
antibody form
fractionated antiserum
clone
polyclonal
technique(s)
Ouchterlony double diffusion: 1:4
storage temp.
2-8°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service