Key Documents
ETIPRQ1250-EP
Eppendorf® epT.I.P.S. standard
(dark green), size, pack of 960 ea (10 trays × 96 tips)
About This Item
UNSPSC Code:
12352200
sterility:
non-sterile
feature:
Eppendorf Quality
packaging:
pack of 960 ea (10 trays × 96 tips)
pack of 960 ea (Autoclavable refill trays)
manufacturer/tradename:
Eppendorf® 022494018 (NA)
Pricing and availability is not currently available.
material
(dark green)
size
L
sterility
non-sterile
feature
Eppendorf Quality
packaging
pack of 960 ea (10 trays × 96 tips)
pack of 960 ea (Autoclavable refill trays)
manufacturer/tradename
Eppendorf® 022494018 (NA)
maximum volume
1.25 mL
General description
Bulk packed in re-sealable bags.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)