EPS001
HAT Activator, CTPB
A potent activator of HAT (histone acetyltransferase)
Synonym(s):
Benzamide, N-[4-chloro-3-(trifluoromethyl)phenyl]-2-ethoxy-6-pentadecyl, CTPB, CTPB (enzyme activator), CTPB (enzyme inhibitor), N-(4-Chloro-3-trifluoromethyl-phenyl)-2-ethoxy-6-pentadecyl-benzamide
About This Item
Recommended Products
Assay
≥97% (H-NMR)
storage condition
desiccated
protect from light
color
white to off-white
solubility
DMSO: soluble
storage temp.
−20°C
InChI
1S/C31H43ClF3NO2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-18-24-19-17-20-28(38-4-2)29(24)30(37)36-25-21-22-27(32)26(23-25)31(33,34)35/h17,19-23H,3-16,18H2,1-2H3,(H,36,37)
InChI key
OYBMVMAXKOGYDC-UHFFFAOYSA-N
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service