Key Documents
EPM1282-9908
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 1 L Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum
About This Item
UNSPSC Code:
41103816
NACRES:
NB.73
Product EPM1282-9908 is not currently sold in your country. Contact Technical Service
material
aluminum
feature
dedicated
for 1 L Erlenmeyer flasks
non-Interchangeable
manufacturer/tradename
Eppendorf® M1282-9908 (Global)
capacity
(15 x 1 L Erlenmeyer flasks)
General description
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)