Skip to Content
Merck
All Photos(1)

Key Documents

EPI017

Sigma-Aldrich

SIRT6 Inhibitor Screening Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

SIRT6 or Sirtuin 6 proteins are a class of proteins that possess either histone deacetylase or mono-ribosyltransferase activity. SIRT6 is a nuclear sirtuin that has been associated with aging, cellular protection, sugar metabolism and certain types of cancer. Broad therapeutic applications are foreseen for SIRT6 inhibitors, including uses in diabetes, immune-mediated disorders, and cancer. Unlike other known protein deacetylases, which simply hydrolyze acetyl-lysine residues, the sirtuin-mediated deacetylation reaction hydrolyzes acetyl-lysine and NAD. This hydrolysis yields the deacetylated substrate, O-acetyl-ADP-ribose and nicotinamide, itself an inhibitor of sirtuin activity. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. 

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for screening, characterizing and studying SIRT6 inhibitors

Principle

In this Sirtuin 6 inhibitor screening kit, Sirtuin 6 deacetylates the substrate, followed by cleavage of the deacetylated substrate to release the fluorescent group, which is detected fluorometrically (λex = 400 nm/λem = 505 nm).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of sirtuin 6 suppresses inflammatory responses and bone destruction in mice with collagen-induced arthritis.
Lee HS, et al.
Arthritis and Rheumatism, 65, 1776-1785 (2013)
SIRT6 inhibitors with salicylate-like structure show immunosuppressive and chemosensitizing effects.
Damonte P, et al.
Bioorganic & Medicinal Chemistry, 25, 5849-5858 (2017)
Sirtuin 6 protects cardiomyocytes from hypertrophy in vitro via inhibition of NF-?B-dependent transcriptional activity.
Yu SS, et al.
British Journal of Pharmacology, 168, 117-128 (2013)
Neural sirtuin 6 (Sirt6) ablation attenuates somatic growth and causes obesity.
Schwer B, et al.
Proceedings of the National Academy of Sciences of the USA, 107, 21790-21794 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service