EP5920737007
Adapter for Eppendorf® S-4x Universal Large Rotor
capacity, MTP, DWP, PCRplates, pack of 2
Synonym(s):
Adapter for Eppendorf Rotor, Eppendorf Rotor Adapter, Large Rotor Adapter
Select a Size
€325.00
Select a Size
About This Item
€325.00
Recommended Products
material
flat bottom
feature
capacity MTP, DWP, PCR
plates
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5920737007 (Global)
parameter
3,938 × g max. RCF
technique(s)
PCR: suitable
centrifugation: suitable
H
116 mm
compatibility
for use with Universal-Large bucket
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service