Key Documents
EP5810743001
Eppendorf® Swing-bucket rotor A-4-81
without buckets
Select a Size
1 EA
€1,530.00
€1,530.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€1,530.00
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
€1,530.00
Please contact Customer Service for Availability
feature
to hold 4 x 4 MTP/flex buckets
to hold 4 x 500 mL rectangular buckets
without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5810743001
parameter
4,000 max. RCF (3,220 xg)
4,000 rpm (3,220 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)