Skip to Content
Merck
All Photos(1)

Key Documents

EP5804775007

Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL

holds 1 tube 50 mL Falcon, pack of 2 ea

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
€97.30

€97.30


Please contact Customer Service for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
2 EA
€97.30

About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

€97.30


Please contact Customer Service for Availability

Notify Me

Get notified when this item is ready to ship via email.

feature

Holds 50 mL tubes
holds 1 tube 50 mL Falcon

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5804775007

technique(s)

centrifugation: suitable

compatibility

for use with Rotor F-34-6-38

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service