Skip to Content
Merck
All Photos(1)

Documents

EP5702741000

Eppendorf® 20 Rubber Mats

for use with 5702 737.003, pack of 20 ea

Synonym(s):

Rubber Mat for Eppendorf® A-4-38 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

feature

Holds 7 mL tubes

packaging

pack of 20 ea

manufacturer/tradename

Eppendorf® 5702741000

technique(s)

centrifugation: suitable

compatibility

for use with 5702 737.003

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service