Key Documents
EP5702718009
Eppendorf® Adapter for 85/100 mL Round Bucket
pack of 2 ea
Select a Size
2 EA
€109.00
€109.00
Please contact Customer Service for Availability
Select a Size
Change View
2 EA
€109.00
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
€109.00
Please contact Customer Service for Availability
feature
Holds 85 - 100 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702718009
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round buckets
General description
adapter for use with Eppendorf 85/100 mL round bucket
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)