Documents
EP5490036000
Rotor for Eppendorf® Concentrator Plus
F-45-24-12, non aerosol tight, to hold, 24 x 6.0/8.0 mL round-bottom tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 24 x 12 x 67 -100 mm
to hold 24 x 6.0/8.0 mL round-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490036000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
Legal Information
Eppendorf is a registered trademark of Eppendorf AG