Skip to Content
Merck
All Photos(1)

Key Documents

EP5427700005

Eppendorf® Rotor A-2-MTP

incl. 2x MTP- buckets and rotor lid

Synonym(s):

A-2-MTP Rotor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,450.00

€1,450.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€1,450.00

About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

€1,450.00


Please contact Customer Service for Availability

feature

to hold 2  x MTP, PCR or deepwell plates (swing-bucket rotor)
to hold 2  x swing-bucket rotor
incl. 2x MTP- buckets and rotor lid

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5427700005

parameter

4,680 max. RCF (2,204 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5430/5430R

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service