Skip to Content
Merck
All Photos(1)

Key Documents

EP4987000398

Eppendorf® Repeater E3/E3x

E3 bundle, with charging cable and Combitips advanced assortment pack (1 Combitip of each size), 1 ul to 50 ml, AC/DC input 100 V / 240 V, 50-60 Hz, US 2-pin plug

Synonym(s):

Adjustable volume pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
Product EP4987000398 is not currently sold in your country. Contact Technical Service

sterility

non-sterile

feature

E3 bundle
with charging cable and Combitips advanced assortment pack (1 Combitip of each size)

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 4987000398 (US)

measuring range

1 ul to 50 ml

maximum volume

50 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Multipette E3/E3x in combination with the Combitips® advanced creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results. The motor driven dispenser gives you full volume control, improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user; filling the tip once allows dispensing up to 100 times. The system is an essential lab tool ideal for all liquid handling applications, accommodating more than 5,000 different dispensing volumes between 1 μL to 50 mL.

Legal Information

Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In this article we provide a complete workflow of quantification and identification of acylcarnitines in dried blood spots (DBS) using stable isotope-labeled (SIL) acylcarnitines CRMs by FIA-MS/MS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service