EP4861000834
Eppendorf® Xplorer® Move It pipette
no. of channels, 6, cone volume × distance 50-1,200 μL × 9 mm, green
Select a Size
€2,130.00
Select a Size
About This Item
€2,130.00
Recommended Products
material
green
feature
no. of channels 6
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4861000834
cone volume × distance
50-1,200 μL × 9 mm
Related Categories
General description
- Adjustable tip spacing pipette
- Multi-channel pipette
- Electronic pipette
- 6-channel
- 50-1,200μL
- 9mm cone distance
- Green
Application
- Automated pipetting of liquids (Pip) for forward pipetting
- Manual pipetting (Man)for volume determination
- Pipetting and mixing (P/M) for automated mixing of samples
- Dispensing (Dis)for manual filling of plates, gels, and reaction vessels
- Automatic dispensing (Ads for automated filling of plates, gels, and reaction vessels
- Multi aspiration: Pooling supernatants from different vessels (SPC)
- Sequential dispensing and pipetting (Spc) for user-defined volume steps
- Reverse pipetting (Spc) for non-aqueous solutions
- Diluting (Spc) for preparation of dilution series
- Fix volume (Fix): Save up to 10 volumes for ease of operation
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)