EP3880000151
Transfer Rack
for Eppendorf® SmartBlock 1.5 mL and Eppendorf® SmartBlock 2.0 mL
Select a Size
€50.60
Select a Size
About This Item
€50.60
Recommended Products
feature
Holds 24 x 1.5-2 mL tubes
Limited to ThermoMixer C annd ThermoStat C
for Eppendorf® SmartBlock 1.5 mL and Eppendorf® SmartBlock 2.0 mL
manufacturer/tradename
Eppendorf® 3880000151 (Global)
parameter
-96 °C temp. range (121 °C)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Holds up to 24 microtubes (1.5 mL or 2.0 mL) for easy transfer of multiple vessels in parallel
- Convenient transfer of up to 24 hot vessels without burning the fingers
- Temperature range from -96 °C to 121 °C for broad flexibility
- Limited to ThermoMixer C annd ThermoStat C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service