Skip to Content
Merck
All Photos(2)

Key Documents

EP3123000942

Sigma-Aldrich

Eppendorf® Research plus Pipettes with Carousel and epT.I.P.S.®

0.1-2.5 / 0.5-10 / 2-20 / 10-100 / 20-200 / 100-1000 µL

Synonym(s):

pipettors

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,650.00

€1,650.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€1,650.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.02

€1,650.00


Please contact Customer Service for Availability

feature

no. of channels 1 x Channel
Mechanical

manufacturer/tradename

Eppendorf® 3123000942

volume

0.1-  2.5 μL
0.5- 10 μL
100 - 1,000 μL
10 - 100 μL
20 - 200 μL
2 - 20 μL

General description

This pipette enables fatigue-free work and intuitive handling. Eppendorf® Research plus pipettes meet the highest standards of precision and accuracy - with ultimate ergonomics and enormous versatility.

Application

Liquid handling

Features and Benefits

  • Mechanical air-cushion pipette for accurate, easy & ergonomic pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: this ultra-light pipette is designed according to the Eppendorf PhysioCare Concept® principles
  • Enjoy minimal tip attachment forces due to a spring-loaded tip cone (available for all pipettes up to 1 mL) that reduces stress without sacrificing tip tightness
  • Benefit from one of the lowest operating and tip ejection forces on the pipette market, helping to reduce hand and arm strain
  • Temporarily adjust your pipette in seconds for better accuracy when pipetting challenging liquids like ethanol or when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs and applications to ensure a proper decontamination
  • Premium material ensures a high UV and chemical resistance

Packaging

0.1-2.5 µL / 0.5-10µL / 2-20 µL (yellow) /10-100 µL / 20-200 µL /100-1000 µL pipettes incl. epT.I.P.S.and pipette carousel 2

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
PhysioCare Concept is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service