Skip to Content
Merck
All Photos(1)

Documents

EP3123000268

Eppendorf® Research plus G, Basic

single-channel, variable, volume 100-1000 μL, blue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

single-channel
variable

packaging

pack of 1 ea
pkg of 1 ea

manufacturer/tradename

Eppendorf® 3123000268

volume

100-1000 μL

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

The Eppendorf Research plus is one of the safest and most ergonomic mechanical pipettes currently available. It not only meets the highest requirements for precision and accuracy, but also offers a high degree of flexibility thanks to a large selection of variants. This makes the Research plus an indispensable companion in your laboratory. Thanks to its remarkably low weight and reduced effort when pipetting, this micropipette helps to relieve the strain on your hand and arm.
The Eppendorf Research plus pipette is designed as a piston-stroke pipette specifically for aspirating and dispensing liquids. It is recognized as one of the safest and most ergonomic mechanical pipettes on the market. With its exceptional precision and accuracy, the Research plus meets the highest standards of performance. Additionally, it offers great flexibility with a wide range of variants available. As a result, the Eppendorf Research plus pipette is a reliable and indispensable companion for laboratory work.

Application

  • Forward pipetting
  • Reverse pipetting
  • Removal of excess material
  • Sample mixing
  • Phase extraction
  • Filling of plates, gels and reaction vessels
Eppendorf Research plus G, Basic is suitable for general laboratory use to transfer liquids in the specified volume range in combination with matching tips. Some of the common applications of this pipette include forward pipetting, reverse pipetting, supernatant removal, sample mixing, phase extraction, as well as filling plates, gels, and reaction vessels.

Features and Benefits

  • Mechanical air cushion pipette with three-button operation as standard (one control for volume selection, one for pipetting and one for tip ejection)
  • Ideal for accurate, easy and ergonomic pipetting of aqueous solutions by hand
  • Convince yourself of the difference in weight and effort required when pipetting: Developed according to the principles of the Eppendorf PhysioCare Concept
  • Benefit from one of the pipettes with the lowest operating and tip ejection forces on the pipette market to reduce hand and arm strain
  • Minimized attachment forces thanks to a spring-loaded tip cone (available for all pipettes up to 1 mL)
  • Adjust your pipette in seconds to improve accuracy when pipetting difficult liquids such as ethanol or when pipetting at high altitudes. Then return to factory adjustment without calibration
  • Depending on your needs and application, autoclave the entire pipette or just the base to ensure proper decontamination
  • High-quality material ensures high UV and chemical resistance; special single-channel bases available for aggressive chemicals
  • Single-channel pipettes with ACT label and epT.IPS pipette tips help you make an informed choice when it comes to sustainability

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service