Key Documents
EP022822225
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-8-17, non aerosol tight, (8 x 15 mL conical tubes)
About This Item
UNSPSC Code:
41103909
Product EP022822225 is not currently sold in your country. Contact Technical Service
feature
to hold 8 x 15 mL conical tubes
to hold 8 x 17 x 118 - 123 mm tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822225 (NA)
technique(s)
centrifugation: suitable
capacity
(8 x 15 mL conical tubes)
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
Related Categories
General description
Selection of rotors for the Eppendorf Vacufuge Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)