Key Documents
EP022822047
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-70-11, non aerosol tight, to hold, 70 x 1.5/2.0 mL reaction vessels
About This Item
UNSPSC Code:
41103909
feature
to hold 70 x 1.5/2.0 mL reaction vessels
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822047 (NA)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG