Key Documents
EP0030129644
Eppendorf twin.tec® Real-Time PCR Plates 96
Semi-Skirted, Forensic DNA Grade, working volume 250 μL, white plate, pack of 10 (individually wrapped)
Select a Size
10 EA
€76.50
€76.50
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
€76.50
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
material:
white plate
white wells
size:
96 well
feature:
skirt
Forensic DNA Grade
Semi-Skirted
€76.50
Please contact Customer Service for Availability
material
white plate
white wells
well size
96
feature
skirt
Forensic DNA Grade
Semi-Skirted
packaging
pack of 10 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030129644 (Global)
technique(s)
PCR: suitable
size
96 well
working volume
250 μL
General description
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Features and Benefits
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)