EP0030128672
Eppendorf® Twin tec PCR plates, 96 wells
blue plate, conical bottom colorless wells, skirted, pkg of 25 ea
Synonym(s):
cell culture plate, multi well plates, multiple well plates
Select a Size
€154.00
Select a Size
About This Item
conical bottom colorless wells
polycarbonate
€154.00
Recommended Products
material
blue plate
conical bottom colorless wells
polycarbonate
sterility
non-sterile
feature
skirt (Skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128672
technique(s)
PCR: suitable
size
96 wells
well maximum volume
150 μm
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Eppendorf® Twin tec PCR plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and extremely robust polycarbonate frames for ultimate rigidity and torque-resistance.
- Optimized rim for barcodes and automated work processes
- Within SBS footprint recommendations (127.76x85.47 mm±0.25 mm).
- Stackable.
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block.
- Low profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes.
- 150 μL max. well volume (when used with cap strips).
- Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service