Skip to Content
Merck
All Photos(1)

Documents

EP0030127854

Eppendorf® heat sealing foil

pkg of 100 ea

Synonym(s):

Heat sealing foil

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.17

sterility

non-sterile

packaging

pkg of 100 ea

manufacturer/tradename

Eppendorf® 0030127854

General description

Achieve reliable sealing of all your PCR plates. Eppendorf® PCR Films and Foils ensure that the seal of your PCR plates is airtight.

Application

Eppendorf® heat sealing foil is suitable for use in automated systems.

Features and Benefits

  • Eppendorf® heat sealing foil is used for continuous locking with the best evaporation protection during PCR
  • It is made of aluminum, can be pierced, and protects light-sensitive samples
  • Easily removable
  • Seal integrity is between –80 °C to 100 °C
  • It takes 4 secs of sealing time with Eppendorf heat sealer

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service