Key Documents
EP0030127749
Eppendorf® Serological Pipets, Sterile Grade
violet, capacity 50 mL, pack of 160 ea (4 bags × 40 pcs.)
Select a Size
160 EA
€290.00
€290.00
Please contact Customer Service for Availability
Select a Size
Change View
160 EA
€290.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
€290.00
Please contact Customer Service for Availability
material
violet
sterility
sterile
feature
DNA free
graduations
plugged
packaging
pack of 160 ea (4 bags × 40 pcs.)
manufacturer/tradename
Eppendorf® 0030127749 (Global)
capacity
50 mL
General description
  • Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
  • Easy-to-read graduations for accurate volume determination.
  • Pipets are color-coded based on sizes for easy identification.
  • Sterility assurance level of 10-6.
  • Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
  • Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.
Application
  • Transfer of liquids
  • Mixing suspensions
  • Combining reagents and chemical solutions
  • Transferring cells for empirical analysis or expansion
  • Layering reagents for creating higher density gradients
Legal Information
Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)