Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122216

Eppendorf® Protein LoBind Tubes

capacity 15 mL, PCR clean, pack of 200 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

200 EA
€106.00

€106.00


Please contact Customer Service for Availability


Select a Size

Change View
200 EA
€106.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

€106.00


Please contact Customer Service for Availability

material

colorless tube
conical bottom
screw top cap
white cap

sterility

non-sterile

feature

PCR clean
graduations
neck thread External thread

packaging

pack of 200 ea

manufacturer/tradename

Eppendorf® 0030122216

technique(s)

PCR: suitable

capacity

15 mL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

Application

  • Preparation or storage of protein, peptide or antibody samples.
  • Enzymatic assays – the hydrophilic surface reduces denaturation when it comes into contact with the tube inner wall.
  • Prevention of sample loss during storage of virus stock solutions.
  • Storage of cell suspensions.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service