Key Documents
EP0030077539
Eppendorf® ep Dualfilter T.I.P.S.®
10 racks x 96 ea, PCR clean, capacity 2-20 μL, sterile
About This Item
UNSPSC Code:
41121600
volume:
2—20 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077539
Pricing and availability is not currently available.
material
yellow (Tray)
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030077539
technique(s)
PCR: suitable
capacity
2-20 μL
maximum volume
20 μL
volume
2—20 μL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® (2-20 μL) are uncoated, free of additives and do not leach into the sample.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile
  • Sterile and pyrogen-free.
  • No Coating.
Application
  • Cell culture (media).
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR.
  • Enzymatic reactions (restriction decomposition, ligation).
  • Isolation and purification of nucleic acid.
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS.
  • Proteomics (all types of protein examinations).
  • Protein isolation and purification.
  • NGS library preparation.
Legal Information
Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)