Skip to Content
Merck
All Photos(1)

Key Documents

EMU094441

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ptprj

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGTGGGTTTGCAGAGGAATATGAGGACCTGAAGCTGATTGGGATAAGTTTACCTAAATACACAGCTGAGATAGCCGAGAACAGAGGGAAGAACCGCTACAACAATGTTCTGCCCTATGATATTTCTCGAGTCAAACTTTCAGTCCAGACCCATTCGACAGATGACTACATCAATGCCAACTATATGCCTGGCTACCATTCCAAGAAAGATTTCATTGCCACACAAGGACCTTTACCCAACACTTTGAAAGATTTCTGGCGTATGGTTTGGGAGAAAAACGTATATGCCATTGTTATGTTGACCAAATGCGTGGAGCAGGGAAGGACCAAATGTGAGGAGTACTGGCCTTCCAAGCAGGCTCAGGACTACGGGGACATAACTGTGGCGATGACATCAGAAGTCGTTCTTCCAGAATGGACC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory C Sartor et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(45), 15062-15072 (2015-11-13)
Epigenetic processes that regulate histone acetylation play an essential role in behavioral and molecular responses to cocaine. To date, however, only a small fraction of the mechanisms involved in the addiction-associated acetylome have been investigated. Members of the bromodomain and
K Spring et al.
Oncogene, 34(44), 5536-5547 (2015-03-17)
DEP-1/PTPRJ is a receptor-like protein tyrosine phosphatase mainly known for its antiproliferative and tumor-suppressive functions. Many identified substrates are growth factor receptors, and DEP-1 is deleted and/or mutated in human cancers including that of the breast. However, DEP-1 was also
Xiaoling Luo et al.
OncoTargets and therapy, 8, 3159-3167 (2015-11-26)
Interaction between microRNA (miR-328) and PTPRJ (protein tyrosine phosphatase, receptor type, J) has been reported to be responsible for miR-328-dependent increase in epithelial cancer cell proliferation. However, the role of miR-328 and PTPRJ in hepatocellular carcinoma (HCC) remains unclear. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service