Skip to Content
Merck
All Photos(1)

Key Documents

EMU078881

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ebna1bp2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCCTCAGATCAAGAGTTGCAGGATGCGTTTTCCCGCGGACTCCTGAAGCCAGGTCTCAATGTCGTGCTAGAGAAGCCGAAGAAGGCAGTGAATGACGTGAGTGGCCTGAAACAGTGCTTGGCTGAATTCAGACGGGATCTGGAGTGGGTTGAAAGGCTCGATGTGACCCTGGGTCCAGTGCCAGAAGTCAGTGAGACTCAGCCAACACCTCAGAACCAGGACCAGAAGAAAGGCGTTAATCCAGAAGACGACTTCCAGAGGGAAATGAGTTTCTACCGGCAGGCCCAGGCAGCAGTGCTTGCGGTATTACCCCGACTCCATCAGCTCCAAGTCCCTACGAAGAGGCCCACCGATTATTTTGCAGAAATGGCCAAGTCTGATCAGCAGATGCAAAAGATTCGACAGAAGCTGCAGACTAAACAGGCCGCCATGGAGAAATCTGAAAAGGCCAAGCAACT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU089631EMU023141EMU038421
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Ensembl | mouse accession no.

ENSMUSG00000028729

Ensembl | mouse accession no.

ENSMUSG00000027305

Ensembl | mouse accession no.

ENSMUSG00000019080

Ensembl | mouse accession no.

ENSMUSG00000028167

esiRNA cDNA target sequence

GGCCTCAGATCAAGAGTTGCAGGATGCGTTTTCCCGCGGACTCCTGAAGCCAGGTCTCAATGTCGTGCTAGAGAAGCCGAAGAAGGCAGTGAATGACGTGAGTGGCCTGAAACAGTGCTTGGCTGAATTCAGACGGGATCTGGAGTGGGTTGAAAGGCTCGATGTGACCCTGGGTCCAGTGCCAGAAGTCAGTGAGACTCAGCCAACACCTCAGAACCAGGACCAGAAGAAAGGCGTTAATCCAGAAGACGACTTCCAGAGGGAAATGAGTTTCTACCGGCAGGCCCAGGCAGCAGTGCTTGCGGTATTACCCCGACTCCATCAGCTCCAAGTCCCTACGAAGAGGCCCACCGATTATTTTGCAGAAATGGCCAAGTCTGATCAGCAGATGCAAAAGATTCGACAGAAGCTGCAGACTAAACAGGCCGCCATGGAGAAATCTGAAAAGGCCAAGCAACT

esiRNA cDNA target sequence

GTGCCATGATCTCCAGGATTCCACGGGGTGCTTTTGAGAGGAAGCTGTCTTATGATTGGTCCCAGTTCAATACTCTGTATCTCCGAGTTCGTGGGGATGGACGGCCTTGGATGGTAAATATCAGGCAAGACACGGAGTTTATCCAGAGAAAAAATCAGATGTACAGTTACTTCATGTTCACCCGTGGGGGACCCTACTGGCAGGAAGTCAAGATTCCTTTCTCCAAATTTTTCTTTTCAAATCAAGGAAGGGTACGAGATGTCCAAGGCCCACTCGTATTGGACAAGATCTCTTCTATAGGATTCACTCTGTCTGATAAGGTGGATGGACCATTCTTCCTGGAAATAGATTTCATTGGTGTGTTTACTGATCCAGCCCATACAGAAGAATTTGCTTATGAGAATTCTCCCGTTCTTAACCCAAGACTCTTCAGATAGAGAACTGGGGACGTCAGTAGAGTAGTGGTCACAAAGCAGACAGAGCCTTTCTGTCTACCCGACCCCTTTCGTAGTACTTACTTAAGACGGTAAGTACAGCATGGCCTCTCAGTG

esiRNA cDNA target sequence

TACAGCTTCCTGGCCACTCTGGAACTGTTGGGCAAGCTGCTCCCAGGTACCCTAGCCGGCGTGCTGGCCGACAGCCTAGGCCCACATCTCTGCTTTGCTGTCTTCCTTGTACTCTCAGCCCTGCCTGTTCTGGATCTCCGCCTGGCACCCAGTAACCTGACCTGAGCTGGATGTTGGTTAGGGCTGTCTATAAAGCCAATAAAGCTATGTATGGCCTGGATGTCTGGGACACAGGAAATGCAGCTCTCCTCAAACTGAAGGCTATGTTACCAGAAAGCACATAAAATTACTGAATGCTTTTTATTCTGCAGTTTGGATCTTCTTATCCCCTTCCTGCAGCCAGAAGGTTTTGCCTGACAGTCAGTGATGGCCACATCATGGCCTTGGTGAGCCTGACACCCTCGGAGTAGTGGCCACAGTCTGGCTTGCCTCATGCCTCAATCATGTG

esiRNA cDNA target sequence

CGACTGGACGGCAAAGTTATTGTCCTGACAGCTGCCGCTCAAGGGATTGGACGGGCATCCGCATTAGCTTTTGCAAGAGAAGGAGCCAAAGTCATAGCCACAGATATCAACGAGTCCAAACTCCAGGAGCTGGAAAGTTACCGAGGTATTCAAACTCGAGTCCTTGATGTCACAAAGAAGAGACAGATTGATCAGTTTGCCTCAGAAATCGAGAGAATTGATGTTCTCTTTAATGTTGCTGGTTTTGTCCACCACGGAACCATCCTGGATTGCGAGGAAAAAGACTGGGACTTCTCAATGAATCTCAACGTCCGCAGCATGTTCCTGATGATCAAAGCATTCCTTCCCAAAATGCTTGCTCAGAAGTCAGGCAACATTATCAACATGTCGTCTGTGGCCTCCAGCATCAAAGGGGTGGAGAACAGATGTGTGTACAGTGCTACCAAGGCGGCTGTGATCGGTCTCACCAAGTCCGTGGCTGCAGACTTCATCCAACAGGGCAT

NCBI accession no.

NM_026932

NCBI accession no.

NM_027175

NCBI accession no.

NM_027122

NCBI accession no.

NP_081484

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service