Skip to Content
Merck
All Photos(1)

Key Documents

EMU076311

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Rela

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCATGTCTCACTCCACAGCTGAGCCCATGCTGATGGAGTACCCTGAAGCTATAACTCGCCTGGTGACAGGGTCCCAGAGGCCCCCTGACCCAGCTCCCACACCCCTGGGGACCTCGGGGCTTCCCAATGGTCTCTCCGGAGATGAAGACTTCTCCTCCATTGCGGACATGGACTTCTCTGCTCTTTTGAGTCAGATCAGCTCCTAAGGTGCTGACAGCGACCCTGCTCAGAGCACCAGGTTTCAGGGCACTGAAGCCTTCCCGAAGTGCGTACACATTCTGGGGAGTGTGCTCCAGCTGCCCCCGACTTGTTTGGGTGATCTCTCTGGGGCGGCACGTTTTACTCTTTATCTCGCTTTCGGAGGTGCTTTCGCAGGAGCATTAACCTCCTGGAGACGGAGCTGGGAGGACTCGGTGCATCCCTGTGTTGATAGCTCCTGCTTCGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Zhang et al.
European review for medical and pharmacological sciences, 18(9), 1361-1367 (2014-05-29)
S100A4 is a member of the S100 family of calcium-binding proteins, which possesses a wide range of biological functions, such as regulation of angiogenesis, cell survival, motility, and invasion. Here, we demonstrate for the first time a major role of
Martyn K White et al.
PloS one, 9(10), e110122-e110122 (2014-10-14)
The human neurotropic polyomavirus JC (JCV) causes the fatal CNS demyelinating disease progressive multifocal leukoencephalopathy (PML). JCV infection is very common and after primary infection, the virus is able to persist in an asymptomatic state. Rarely, and usually only under
Yun Qu et al.
Cell biochemistry and function, 33(5), 320-325 (2015-07-17)
Nuclear factor-kappaB (NF-κB) is an important transcriptional factor and regulates a variety of pathophysiologic process involved in cell survival and death. This study aimed to assess the effects of NF-κB p65 subunit knockdown in suppression of nude mouse lung tumour
Maroof Alam et al.
Oncotarget, 5(9), 2622-2634 (2014-04-29)
The capacity of breast cancer cells to form mammospheres in non-adherent serum-free culture is used as a functional characteristic of the self-renewing stem-like cell population. The present studies demonstrate that silencing expression of the MUC1-C oncoprotein inhibits growth of luminal
Yuan Zhang et al.
Journal of neuroinflammation, 12, 156-156 (2015-09-05)
Mounting evidence has indicated that high-mobility group box 1 (HMGB1) is involved in cell activation and migration. Our previous study demonstrated that methamphetamine mediates activation of astrocytes via sigma-1 receptor (σ-1R). However, the elements downstream of σ-1R in this process

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service