Skip to Content
Merck
All Photos(1)

Key Documents

EMU066761

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Fyb

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGCCTCAGACTTCCCTCCTCCACCTGCAGAGATGAGTCAAGGAATGAGTGTTGGAAGGGCAAAAACGGAAGAGAAAGATCCCAAGAAGCTAAAAAAGCAAGAAAAGGAAGAAAAAGACCTCAGGAAAAAATTTAAGTACGACGGTGAAATTCGAGTTCTATATTCAACTAAAGTTGCGTCCTCCTTAACCTCTAAAAAGTGGGGAGCGAGAGATCTGCAGATAAAACCTGGGGAGTCACTCGAAGTTATACAAAGCACAGATGACACCAAAGTTCTCTGCAGGAATGAAGAGGGCAAATATGGTTATGTCCTTCGGAGTTACCTGGTGGACAATGATGGAGAAATCTATGACGACATCGCTGATGGTTGCATCTATGACAATGACTAGCACTCTGCTCTGTTCATTCCACTGTGCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Wang et al.
Folia histochemica et cytobiologica, 58(2), 96-107 (2020-06-27)
Growing evidence indicates that Rictor (Rapamycin-insensitive companion of mTOR) is overexpressed across several malignancies and associated with poor survival. However, only limited data indicate that Rictor plays a role in gastric cancer (GC). We sought to explore the prognostic value
Mahmoud A Chawsheen et al.
Cellular signalling, 81, 109934-109934 (2021-02-06)
Lung cancer has a poor prognosis partly due to a lack of response to treatments such as the chemotherapy drug gemcitabine. Combinations of chemotherapy drugs with signal transduction inhibitors may be more effective treatments. In this study we have investigated
Zhaoming Lu et al.
Acta pharmaceutica Sinica. B, 10(6), 1004-1019 (2020-07-10)
Dysregulation of mTORC1/mTORC2 pathway is observed in many cancers and mTORC1 inhibitors have been used clinically in many tumor types; however, the mechanism of mTORC2 in tumorigenesis is still obscure. Here, we mainly explored the potential role of mTORC2 in
Li Feng et al.
Pharmaceutical biology, 57(1), 586-594 (2019-09-08)
Context: Evidence suggests that microRNA (miRNA) regulate gene expression and bone tissue homoeostasis of osteoporosis. MiR-152 has found to be abnormally expressed in osteoporosis, but its role in osteoblast differentiation has not been elucidated. Objective: To understand the potential mechanism
Thomas Jansson et al.
Clinical science (London, England : 1979), 134(1), 53-70 (2019-12-12)
Intrauterine growth restriction (IUGR) increases the risk for perinatal complications and metabolic and cardiovascular disease later in life. The syncytiotrophoblast (ST) is the transporting epithelium of the human placenta, and decreased expression of amino acid transporter isoforms in the ST

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service