Skip to Content
Merck
All Photos(1)

Key Documents

EMU057611

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Notch1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCAACCCATGTCAGAATGATGCCACTTGCCTGGACCAGATTGGGGAGTTCCAATGCATATGTATGCCAGGTTATGAAGGTGTATACTGTGAAATCAACACGGATGAGTGCGCCAGCAGCCCCTGTCTGCACAATGGCCACTGCATGGACAAGATCAATGAGTTCCAATGTCAGTGCCCCAAAGGCTTCAACGGGCACCTGTGCCAGTATGATGTGGATGAGTGTGCCAGCACACCATGCAAGAACGGTGCCAAGTGCCTGGATGGGCCCAACACCTATACCTGCGTGTGTACAGAAGGTTACACAGGGACCCACTGCGAAGTGGACATTGACGAGTGTGACCCTGACCCCTGCCACTATGGTTCCTGTAAGGATGGTGTGGCCACCTTTACCTGCCTGTGCCAGCCAGGCTACACAGGCCATCACTGTGAGACCAACATCAATGAGTGCCACAGCCAACCGTGCCGCCATGGGGGCACCTGCCAGGACCGTGACAACTCCTACCTCTGCTTATGCCTCAAGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debarshi Banerjee et al.
Cancer research, 75(8), 1592-1602 (2015-03-07)
The Notch pathway plays multiple key roles in tumorigenesis, and its signaling components have therefore aroused great interest as targets for emerging therapies. Here, we show that inhibition of Notch, using a soluble receptor Notch1 decoy, unexpectedly caused a remarkable
Elisabetta Palazzo et al.
International journal of molecular sciences, 16(11), 26291-26302 (2015-11-06)
The Notch signaling pathway orchestrates cell fate by either inducing cell differentiation or maintaining cells in an undifferentiated state. This study aims to evaluate Notch expression and function in normal human keratinocytes. Notch1 is expressed in all epidermal layers, though
Yinan Liu et al.
PloS one, 9(10), e109588-e109588 (2014-10-15)
The Notch signaling pathway plays versatile roles during heart development. However, there is contradictory evidence that Notch pathway either facilitates or impairs cardiomyogenesis in vitro. In this study, we developed iPSCs by reprogramming of murine fibroblasts with GFP expression governed
Yao Cheng et al.
Anti-cancer drugs, 25(7), 778-789 (2014-03-19)
Tumor invasion and migration obstructs the treatment and prognosis of cancer. In this research, we investigated the effect of oroxylin A, a natural compound extracted from Scutellaria radix, the root of Scutellaria baicalensis, on inhibition of the invasion and migration
Guanqiao Wang et al.
Cellular signalling, 27(7), 1369-1379 (2015-04-07)
Carbonic anhydrase IX(CA9)is a member of the carbonic anhydrase family that catalyzes the reversible hydration of carbon dioxide, and plays a key role in the regulation of pH. Although a large number of studies have shown that CA9 is strongly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service