Skip to Content
Merck
All Photos(1)

Key Documents

EMU052411

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Cdc42

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGTTGGTGATGGTGCTGTTGGTAAAACATGTCTCCTGATATCCTACACAACAAACAAATTCCCATCGGAATATGTACCAACTGTTTTTGACAACTATGCAGTCACAGTTATGATTGGTGGAGAGCCATACACTCTTGGACTTTTTGATACTGCAGGGCAAGAGGATTATGACAGACTACGACCGCTAAGTTATCCACAGACAGATGTTTTTCTAGTATGTTTCTCAGTGGTCTCTCCATCCTCATTTGAAAATGTGAAAGAAAAGTGGGTGCCTGAGATAACTCACCACTGTCCAAAGACTCCTTTCTTGCTTGTTGGGACCCAAATTGATCTCAGAGATGACCCCTCTACTATTGAGAAACTTGCCAAGAACAAACAGAAGCCTATTACTCCAGAGACTGCTGAAAAGCTGGCGCGGGATCTGAAGGCTGTCAAGTATGTGGAGTGCTCCGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent Pieuchot et al.
Nature communications, 9(1), 3995-3995 (2018-09-30)
Cells have evolved multiple mechanisms to apprehend and adapt finely to their environment. Here we report a new cellular ability, which we term "curvotaxis" that enables the cells to respond to cell-scale curvature variations, a ubiquitous trait of cellular biotopes.
Shayoni Ray et al.
Molecular biology of the cell, 25(16), 2393-2407 (2014-06-27)
Coordinated actin microfilament and microtubule dynamics is required for salivary gland development, although the mechanisms by which they contribute to branching morphogenesis are not defined. Because LIM kinase (LIMK) regulates both actin and microtubule organization, we investigated the role of
Tilman D Rachner et al.
Breast cancer research : BCR, 16(1), R20-R20 (2014-02-18)
Amino-bisphosphonates and statins inhibit the mevalonate pathway, and may exert anti-tumor effects. The Wnt inhibitor dickkopf-1 (DKK-1) promotes osteolytic bone lesions by inhibiting osteoblast functions and has been implicated as an adverse marker in multiple cancers. We assessed the effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service