Skip to Content
Merck
All Photos(1)

Key Documents

EMU052231

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Park7

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGCAGTGTAGCCGTGATGTAATGATTTGTCCAGATACCAGTCTGGAAGATGCAAAAACGCAGGGACCATACGATGTGGTGGTTCTTCCAGGAGGAAATCTGGGTGCACAGAATTTATCTGAGTCGCCTATGGTGAAGGAGATCCTCAAGGAGCAGGAGAGCAGGAAGGGCCTCATAGCTGCCATCTGTGCAGGTCCTACGGCTCTGTTGGCTCACGAAGTAGGTTTTGGATGCAAGGTCACAACACACCCACTGGCTAAGGACAAAATGATGAATGGCAGTCACTACAGCTACTCAGAGAGCCGCGTGGAGAAGGACGGCCTGATCCTCACCAGCCGCGGGCCGGGGACCAGCTTTGAGTTTGCACTAGCCATTGTGGAGGCACTCGTGGGGAAAGACATGGCCAACCAAGTGAAGGCACCGCTTGTTCTCAAAGACTAGAGCCCAAGCCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung-Min Kim et al.
Scientific reports, 4, 4805-4805 (2014-06-14)
Adipose tissue functions as an endocrine organ, and the development of systemic inflammation in adipose tissue is closely associated with metabolic diseases, such as obesity and insulin resistance. Accordingly, the fine regulation of the inflammatory response caused by obesity has
Ismail Ahmed Ismail et al.
Journal of cellular physiology, 230(9), 2262-2269 (2015-02-14)
2'-Benzoyloxycinnamaldehyde (BCA) is a promising antitumor agent. BCA effectively inhibited proliferation of MDA-MB-435 more than in MCF-7 breast cancer cells. Our recent findings showed that DJ-1 protects MCF7 cells from BCA-induced oxidative stress via its mitochondrial translocation and inhibition of
Hong Zhu et al.
Free radical biology & medicine, 71, 121-132 (2014-04-01)
Dihydroartemisinin (DHA), one of the main metabolites of artemisinin and its derivatives, presents anti-cancer potential in vitro and in vivo. To explore the mechanisms of resistance toward DHA, a DHA-resistant cell line, HeLa/DHA, was established with a resistance factor of
Min Sik Choi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(45), 15123-15131 (2014-11-08)
Emerging evidence suggests that oxidative/nitrosative stress, as occurs during aging, contributes to the pathogenesis of Parkinson's disease (PD). In contrast, detoxification of reactive oxygen species and reactive nitrogen species can protect neurons. DJ-1 has been identified as one of several
Cailing Liu et al.
Investigative ophthalmology & visual science, 55(9), 5551-5560 (2014-08-02)
To investigate the role of DJ-1 in Nrf2-regulated antioxidant defense in corneal endothelial cells (CECs) at baseline and in response to ultraviolet A (UV-A)-induced oxidative stress. DJ-1-deficient CECs were obtained by transfection of an immortalized normal human corneal endothelial cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service