Skip to Content
Merck
All Photos(1)

Key Documents

EMU044991

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Trp63

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCAGACCATCTCTTTTCCACCCCGTGACGAGTGGAATGATTTCAACTTTGACATGGATTCTCGTCGCAACAAGCAGCAGCGTATCAAAGAGGAAGGAGAATGAGCGCCCATTGCGGGGTTCTTCCTGTCTTCTTCCACCTCCCAGCCCCTACAGGGCACGCCTGCTTGATCCTCAGAGCCTTCTCGTTAGCTCTTCTCCTTCTCCTTCTCAGTCTGGTTTCTAAAGGGACGGAGAATTAGGAGGCTGCCTGTTACCTAAAGTCTGACCTGTCACCTGATTCTGATTCTGGCTTTAAGCCTTCAATACTCTTGCTTGCAAGATGCATTGACATTGCTAGATAGAAGTTAGCAAAGAAGCAGTAGGTCTCTTTAAGCCAGTGGAGATCTCTCATTGACTTTTATAAAGCATTTTCAGCCTTATAGTCTAAGACTATATATATAAATATATAAATATCCGATATATATTTTGGGTGTGGGGGGTATTGAGTATTGTTTAAATGTAATTTAATGGAAATTGGAGTTGCACTTATCATCCTTCTTTGGAATTTGCTTGTTTCGGATGGCTGAGCTGTACTCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Ye et al.
International journal of oncology, 44(6), 2153-2159 (2014-04-11)
p63 is a member of the p53 protein family and plays a crucial role in epithelial development. p63 is expressed in many types of tumors including esophageal cancer; however, its function in cancer is controversial and its role in esophageal
Renata L Linardi et al.
Veterinary dermatology, 26(4), 213-e47-213-e47 (2015-05-13)
The limited characterization of equine skin, eye and hoof epithelial stem cell (ESC) and differentiation markers impedes the investigation of the physiology and pathophysiology of these tissues. To characterize ESC and differentiation marker expression in epithelial tissues of the equine
Hulda R Jonsdottir et al.
Laboratory investigation; a journal of technical methods and pathology, 95(12), 1418-1428 (2015-09-22)
Idiopathic pulmonary fibrosis (IPF) is a progressive interstitial lung disease with high morbidity and mortality. The cellular source of the fibrotic process is currently under debate with one suggested mechanism being epithelial-to-mesenchymal transition (EMT) in the alveolar region. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service